Search
Search

Kif5b| Gene Symbol: Kif5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008448.3 | NP_032474.2 | 10, 11 | 1387 | ||
| GenBank mRNA | AK085221.1 | - | 2, 3 | 213 | ||
| AK133535.1 | - | 10, 11 | 1384 | |||
| AK136307.1 | - | 10, 11 | 1384 | |||
| BC013248.1 | - | 10, 11 | 1343 | |||
| BC013817.1 | - | 10, 11 | 1353 | |||
| BC025864.1 | - | 10, 11 | 1356 | |||
| BC040800.1 | - | 10, 11 | 1377 | |||
| BC069920.1 | - | 10, 11 | 1363 | |||
| BC090841.1 | - | 10, 11 | 1363 | 30543821 | ||
| L27153.1 | - | 10, 11 | 981 | |||
| U86090.1 | - | 10, 11 | 1214 | |||