Search
Search

Map7| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198635.1 | NP_001185564.1 | 3 | 327 | ||
| NM_008635.2 | NP_032661.2 | 3 | 327 | |||
| GenBank mRNA | AB098611.1 | - | 3 | 233 | ||
| AK008018.1 | - | 3 | 271 | |||
| AK008182.1 | - | 3 | 250 | |||
| AK014575.1 | - | 3 | 244 | |||
| AK133267.1 | - | 3 | 776 | |||
| BC042771.1 | - | 3 | 252 | |||
| BC044733.1 | - | 3 | 252 | |||
| BC046555.1 | - | 3 | 252 | |||
| BC052637.1 | - | 3 | 263 | 6467153 | ||
| BY277244.1 | - | 3 | 328 | |||
| CJ074623.1 | - | 3 | 304 | |||
| Y15197.1 | - | 3 | 233 | |||