Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAP2K1 SIRNA Assays ›
          Gene Symbol:
          Map2k1
          Gene Name:
          mitogen-activated protein kinase kinase 1
          Gene Aliases:
          MAPKK1, MEKK1, Mek1, Prkmk1
          Chromosome Location:
          Chr.9: 64185770 - 64253605 on Build GRCm38
          UniGene ID:
          Mm.248907
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 63974

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Map2k1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008927.3 NP_032953.1 9 1326
          GenBank mRNA AB041567.1 - 9 1306
          AK146233.1 - 9 1316
          AK146931.1 - 9 1319
          AK154793.1 - 9 1353
          AK165664.1 - 8 860
          AK165894.1 - 9 1327
          AK167336.1 - 9 1293
          AK192210.1 - 4 378
          BC051137.1 - 9 1145
          BC054754.1 - 9 1105 6411664
          L02526.1 - 9 1053

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          regulation of protein phosphorylation
          regulation of vascular smooth muscle contraction
          protein phosphorylation
          response to oxidative stress
          cell cycle arrest
          mitotic nuclear division
          cell proliferation
          negative regulation of cell proliferation
          positive regulation of gene expression
          phosphorylation
          neuron differentiation
          keratinocyte differentiation
          positive regulation of cell migration
          melanosome transport
          regulation of stress-activated MAPK cascade
          positive regulation of transcription elongation from RNA polymerase II promoter
          negative regulation of homotypic cell-cell adhesion
          peptidyl-tyrosine dephosphorylation
          positive regulation of GTPase activity
          positive regulation of cell differentiation
          positive regulation of Ras protein signal transduction
          vesicle transport along microtubule
          Golgi inheritance
          response to axon injury
          regulation of axon regeneration
          neuron projection morphogenesis
          cell motility
          positive regulation of axonogenesis
          protein heterooligomerization
          response to glucocorticoid
          lung morphogenesis
          trachea formation
          epithelial cell proliferation involved in lung morphogenesis
          placenta blood vessel development
          labyrinthine layer development
          ERK1 and ERK2 cascade
          positive regulation of protein serine/threonine kinase activity
          regulation of Golgi inheritance
          cellular senescence
          mitophagy in response to mitochondrial depolarization
          regulation of early endosome to late endosome transport
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein tyrosine kinase activity
          receptor signaling protein tyrosine phosphatase activity
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          Ras GTPase binding
          protein kinase binding
          mitogen-activated protein kinase kinase kinase binding
          protein serine/threonine kinase activator activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-pc55z:80/100.66.76.57:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0