Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFBR2 SIRNA Assays ›
          Gene Symbol:
          Tgfbr2
          Gene Name:
          transforming growth factor, beta receptor II
          Gene Aliases:
          1110020H15Rik, AU042018, DNIIR, RIIDN, TBR-II, TbetaR-II, TbetaRII
          Chromosome Location:
          Chr.9: 116087695 - 116175363 on Build GRCm38
          UniGene ID:
          Mm.172346
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 64989

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tgfbr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009371.3 NP_033397.3 4 781
          NM_029575.3 NP_083851.3 3 706
          GenBank mRNA AF406755.1 - 3 380
          AK043619.1 - 4 865
          AK046561.1 - 4 707
          AK053793.1 - 4 782
          AK148167.1 - 3 706
          AK159327.1 - 4 761
          BC052629.1 - 683
          S69114.1 - 3 417

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031132, L09653, S67770, GU067683.

          Panther Classification:

          Molecular Function -

          serine/threonine protein kinase receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          vasculogenesis
          in utero embryonic development
          positive regulation of mesenchymal cell proliferation
          lens development in camera-type eye
          positive regulation of tolerance induction to self antigen
          positive regulation of B cell tolerance induction
          positive regulation of T cell tolerance induction
          growth plate cartilage development
          growth plate cartilage chondrocyte growth
          protein phosphorylation
          receptor-mediated endocytosis
          apoptotic process
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          Notch signaling pathway
          smoothened signaling pathway
          gastrulation
          brain development
          heart development
          negative regulation of cell proliferation
          organ morphogenesis
          regulation of gene expression
          positive regulation of epithelial cell migration
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cell differentiation
          lung development
          activation of protein kinase activity
          embryonic hemopoiesis
          regulation of growth
          regulation of cell proliferation
          response to drug
          myeloid dendritic cell differentiation
          positive regulation of angiogenesis
          positive regulation of smooth muscle cell proliferation
          embryonic cranial skeleton morphogenesis
          positive regulation of NK T cell differentiation
          cartilage development
          palate development
          negative regulation of cardiac muscle cell proliferation
          pathway-restricted SMAD protein phosphorylation
          lung morphogenesis
          bronchus development
          bronchus morphogenesis
          trachea morphogenesis
          trachea formation
          mammary gland morphogenesis
          lung lobe morphogenesis
          response to cholesterol
          lens fiber cell apoptotic process
          positive regulation of reactive oxygen species metabolic process
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          receptor activity
          transforming growth factor beta-activated receptor activity
          transforming growth factor beta receptor activity, type II
          protein binding
          ATP binding
          glycosaminoglycan binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          mitogen-activated protein kinase kinase kinase binding
          type I transforming growth factor beta receptor binding
          type III transforming growth factor beta receptor binding
          SMAD binding
          metal ion binding
          transforming growth factor beta binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0