Search
Search

Tgfb3| Gene Symbol: Tgfb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009368.3 | NP_033394.2 | 2 | 1507 | ||
| GenBank mRNA | AK132519.1 | - | 2 | 1497 | ||
| AK145304.1 | - | 2 | 1492 | |||
| AK148385.1 | - | 2 | 1371 | |||
| AK162560.1 | - | 2 | 271 | |||
| BC005513.1 | - | 2 | 249 | |||
| BC014690.1 | - | 2 | 786 | 3663150 | ||
| BC089347.1 | - | 2 | 1464 | |||
| BC094591.1 | - | 3 | 932 | |||
| BC108426.1 | - | 2 | 1507 | 6757707 | ||
| M32745.1 | - | 2 | 1002 | |||