Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDC42 SIRNA Assays ›
          Gene Symbol:
          Cdc42
          Gene Name:
          cell division cycle 42
          Gene Aliases:
          AI747189, AU018915
          Chromosome Location:
          Chr.4: 137319696 - 137357759 on Build GRCm38
          UniGene ID:
          Mm.1022
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 66117

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cdc42
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243769.1 NP_001230698.1 4 358
          NM_009861.3 NP_033991.1 4 358
          GenBank mRNA AK003098.1 - 4 279
          AK051543.1 - 4 279
          AK075567.1 - 4 277
          AK144216.1 - 4 281
          AK145659.1 - 3 122
          AK151087.1 - 4 303
          AK151726.1 - 4 280
          AK153564.1 - 4 302
          AK154870.1 - 4 277
          AK159470.1 - 4 320
          AK166281.1 - 4 277
          AK167195.1 - 4 266
          AK167400.1 - 4 271
          AK167609.1 - 4 276
          AK168013.1 - 3 271
          AK168076.1 - 4 271
          AK168089.1 - 3 271
          AK168276.1 - 4 271
          AK168503.1 - 4 292
          AK168758.1 - 4 311
          AK168820.1 - 4 284
          AK169122.1 - 4 292
          AK169232.1 - 4 271
          AK169805.1 - 4 296
          AK213611.1 - 3 132
          CD551112.1 - 4 345
          L11318.1 - 3 194
          L78075.1 - 3 198
          U37720.1 - 4 266

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_171994, XM_008764286, XM_008764287, FQ231846, FQ232179, and others.

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          sprouting angiogenesis
          cardiac conduction system development
          keratinocyte development
          endocytosis
          actin filament organization
          Golgi organization
          regulation of mitotic nuclear division
          nuclear migration
          establishment or maintenance of cell polarity
          small GTPase mediated signal transduction
          Rho protein signal transduction
          nervous system development
          positive regulation of gene expression
          negative regulation of gene expression
          endosomal transport
          single organismal cell-cell adhesion
          cell differentiation
          hair follicle morphogenesis
          keratinization
          regulation of protein stability
          positive regulation of peptidyl-serine phosphorylation
          adherens junction organization
          cellular protein localization
          establishment or maintenance of apical/basal cell polarity
          multicellular organism growth
          dendritic cell migration
          regulation of protein catabolic process
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          regulation of protein heterodimerization activity
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of DNA replication
          regulation of protein kinase activity
          positive regulation of JNK cascade
          filopodium assembly
          neuron fate determination
          epidermis morphogenesis
          actin filament bundle assembly
          regulation of protein metabolic process
          nucleus localization
          establishment of Golgi localization
          positive regulation of synapse structural plasticity
          heart contraction
          canonical Wnt signaling pathway
          epithelial-mesenchymal cell signaling
          hair follicle placode formation
          positive regulation of hair follicle cell proliferation
          organelle transport along microtubule
          actin filament branching
          epithelial cell-cell adhesion
          positive regulation of intracellular protein transport
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          GTP-dependent protein binding
          mitogen-activated protein kinase kinase kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0