Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLN3 SIRNA Assays ›
          Gene Symbol:
          Cln3
          Gene Name:
          ceroid lipofuscinosis, neuronal 3, juvenile (Batten, Spielmeyer-Vogt disease)
          Gene Aliases:
          AI323623
          Chromosome Location:
          Chr.7: 126571400 - 126585817 on Build GRCm38
          UniGene ID:
          Mm.268930
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 66336

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cln3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146311.1 NP_001139783.1 12, 13 1132
          NM_009907.3 NP_034037.3 10, 11 1020
          GenBank mRNA AK078976.1 - 10, 11 1004
          AK134036.1 - 13, 14 1655
          AK184160.1 - 2, 3 83
          AK217815.1 - 2, 3 82
          BC058753.1 - 11, 12 956 6477909
          BC060306.1 - 9, 10 902 5250894
          BC080759.1 - 11, 12 914 5353776
          U47106.1 - 10, 11 970
          U68064.1 - 10, 11 990
          U92811.1 - 10, 11 939

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_000086, NM_001042432, BC111068, AF015598, BC002394, and others.
          This siRNA also targets the following transcripts in Rat: NM_001006971, XM_008759874, XM_008759871, XM_008759872, XM_008759875, and others.

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          globoside metabolic process
          positive regulation of protein phosphorylation
          cellular amino acid metabolic process
          ceramide metabolic process
          glucosylceramide metabolic process
          galactosylceramide metabolic process
          sphingomyelin metabolic process
          transport
          amino acid transport
          endocytosis
          receptor-mediated endocytosis
          vacuolar transport
          lysosome organization
          lysosomal lumen acidification
          associative learning
          regulation of gene expression
          arginine transport
          macroautophagy
          negative regulation of macroautophagy
          cell migration
          protein processing
          actin cytoskeleton organization
          ionotropic glutamate receptor signaling pathway
          lysosomal lumen pH elevation
          neurotransmitter metabolic process
          amyloid precursor protein catabolic process
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          negative regulation of neuron apoptotic process
          negative regulation of proteolysis
          vesicle transport along microtubule
          neuromuscular process controlling balance
          regulation of cytosolic calcium ion concentration
          regulation of filopodium assembly
          membrane organization
          protein localization to membrane
          protein localization to plasma membrane
          autophagosome maturation
          protein binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0