Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSN2 SIRNA Assays ›
Gene Symbol:
Csn2
Gene Name:
casein beta
Gene Aliases:
Csnb
Chromosome Location:
Chr.5: 87692619 - 87699429 on Build GRCm38
UniGene ID:
Mm.268737
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
66477
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Csn2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286020.1
NP_001272949.1
8
763
NM_001286021.1
NP_001272950.1
8
778
NM_001286022.1
NP_001272951.1
7
739
NM_001286023.1
NP_001272952.1
6
721
NM_001286024.1
NP_001272953.1
6
718
NM_009972.2
NP_034102.1
8
766
GenBank mRNA
AK021324.1
-
8
760
AK021328.1
-
8
760
AK052803.1
-
8
763
AK052805.1
-
8
763
AK052822.1
-
8
760
AK085678.1
-
8
760
AK085694.1
-
8
763
AK085698.1
-
8
760
AK085729.1
-
8
763
AK085756.1
-
8
766
AK142616.1
-
8
763
AK142620.1
-
8
763
AK142631.1
-
8
763
AK142635.1
-
8
763
AK164747.1
-
8
763
AK164754.1
-
2
2712
AK164761.1
-
8
763
AK164791.1
-
8
763
AK164792.1
-
8
760
AK164793.1
-
8
760
AK164794.1
-
8
763
BC003863.1
-
3
588
BC003976.1
-
4
612
BC013332.1
-
8
763
3670912
BC019114.1
-
8
744
3483918
BC019189.1
-
7
756
5028194
BC020356.1
-
8
743
BC020383.1
-
8
743
BC021153.1
-
8
761
3670372
BC050270.1
-
8
743
BC057671.1
-
5
2555
BC080709.1
-
8
747
3671502
BC092098.1
-
9
651
X04490.1
-
8
762
Back To Top
More Information
Panther Classification:
Molecular Function -
storage protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
calcium ion transport
lactation
negative regulation of catalytic activity
negative regulation of lactation
enzyme inhibitor activity
transporter activity
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0