Search
Search

Ext2| Gene Symbol: Ext2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010163.3 | NP_034293.2 | 2, 3 | 756 | ||
| GenBank mRNA | AK082572.1 | - | 2, 3 | 738 | ||
| AK148090.1 | - | 2, 3 | 745 | |||
| AK148369.1 | - | 1, 2 | 517 | |||
| AK150155.1 | - | 2, 3 | 748 | |||
| AK152265.1 | - | 856 | ||||
| AK152840.1 | - | 858 | ||||
| AK164342.1 | - | 2, 3 | 742 | |||
| BC006597.1 | - | 2, 3 | 729 | 3965079 | ||
| U67837.1 | - | 2, 3 | 731 | |||
| U72141.1 | - | 2, 3 | 735 | |||