Search
Search

Dync1i1| Gene Symbol: Dync1i1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191023.1 | NP_001177952.1 | 7 | 909 | ||
| NM_001191025.1 | NP_001177954.1 | 7 | 858 | |||
| NM_001191026.1 | NP_001177955.1 | 8 | 885 | |||
| NM_001191027.1 | NP_001177956.1 | 7 | 825 | |||
| NM_010063.4 | NP_034193.2 | 8 | 918 | |||
| GenBank mRNA | AF063229.1 | - | 8 | 752 | ||
| AF063230.1 | - | 7 | 692 | |||
| AK158589.1 | - | 8 | 886 | |||
| BC051992.1 | - | 8 | 646 | 5696629 | ||
| GU992206.1 | - | 7 | 650 | |||
| GU992207.1 | - | 7 | 566 | |||