Search
Search

Gli1| Gene Symbol: Gli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010296.2 | NP_034426.2 | 3, 4 | 269 | ||
| GenBank mRNA | AB025922.1 | - | 3, 4 | 279 | ||
| AB232673.1 | - | 2, 3 | 200 | |||
| AB232674.1 | - | 3, 4 | 296 | |||
| AB232675.1 | - | 3, 4 | 429 | |||
| AB234616.1 | - | 4, 5 | 780 | |||
| AB234617.1 | - | 3, 4 | 661 | |||
| AF026305.1 | - | 3, 4 | 269 | |||
| AK157048.1 | - | 3, 4 | 934 | |||
| BC131649.1 | - | 2, 3 | 247 | |||
| BC131650.1 | - | 2, 3 | 247 | 40110429 | ||
| CJ062804.1 | - | 2, 3 | 253 | |||