Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPSA SIRNA Assays ›
          Gene Symbol:
          Rpsa
          Gene Name:
          ribosomal protein SA
          Gene Aliases:
          67kDa, 67lr, AL022858, Lamr, Lamr1, Lamrl1, MLR, P40, P40-3, P40-8
          Chromosome Location:
          Chr.9: 120127766 - 120132369 on Build GRCm38
          UniGene ID:
          Mm.311972
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68484

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpsa
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011029.4 NP_035159.3 2, 3 228
          GenBank mRNA AF140348.1 - 2, 3 215
          AK010423.1 - 2, 3 208
          AK010985.1 - 2, 3 210
          AK011041.1 - 2, 3 210
          AK011043.1 - 2, 3 210
          AK082935.1 - 2, 3 207
          AK088954.1 - 2, 3 208
          AK134224.1 - 2, 3 208
          AK135488.1 - 2, 3 210
          AK145713.1 - 2, 3 205
          AK151123.1 - 2, 3 207
          AK160551.1 - 2, 3 207
          AK160625.1 - 2, 3 207
          AK165219.1 - 2, 3 208
          AK166697.1 - 2, 3 207
          AK166874.1 - 2, 3 210
          AK167132.1 - 2, 3 208
          AK205896.1 - 1, 2 113
          AK211265.1 - 1, 2 113
          AK212093.1 - 1, 2 113
          AK212679.1 - 1, 2 113
          AK213565.1 - 1, 2 113
          AK214711.1 - 1, 2 113
          AK215661.1 - 1, 2 113
          AK216125.1 - 1, 2 113
          AK218136.1 - 1, 2 113
          AK219743.1 - 1, 2 113
          AK219820.1 - 1, 2 113
          AV586849.1 - 2, 3 305
          BC003829.1 - 2, 3 192 3600738
          BC037195.1 - 1, 2 155 4500748
          BC055886.1 - 1, 2 362 4240083
          BC081461.1 - 1, 2 163 6820702
          BC084677.1 - 2, 3 192 6476344
          BC092041.1 - 2, 3 193 2651126
          BC094902.1 - 2, 3 192 6433349
          BC099601.1 - 2, 3 193 1245698
          BC110285.1 - 2, 3 193 5151374
          EU554633.1 - 1, 2 130
          J02870.1 - 2, 3 209
          X06406.1 - 2, 3 207

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017138, XM_008766672, FQ220822, FQ227056, FQ228002, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          endonucleolytic cleavage in ITS1 to separate SSU-rRNA from 5.8S rRNA and LSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          endonucleolytic cleavage to generate mature 3'-end of SSU-rRNA from (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
          rRNA export from nucleus
          translation
          single organismal cell-cell adhesion
          structural constituent of ribosome
          laminin receptor activity
          ribosome binding
          laminin binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0