Search
Search

Cdk14| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011074.2 | NP_035204.2 | 3 | 358 | ||
| GenBank mRNA | AF033655.1 | - | 3 | 317 | ||
| AK016370.1 | - | 3 | 300 | |||
| AK045083.1 | - | 3 | 359 | |||
| AK051283.1 | - | 3 | 332 | |||
| AK051411.1 | - | 3 | 301 | |||
| AK083269.1 | - | 2 | 513 | |||
| AK083790.1 | - | 3 | 301 | |||
| AK087398.1 | - | 2 | 129 | |||
| AK129226.1 | - | 3 | 185 | |||
| AK138437.1 | - | 2 | 599 | |||
| AK139386.1 | - | 2 | 519 | |||
| AK141535.1 | - | 2 | 287 | |||
| AK165149.1 | - | 2 | 203 | |||
| BC068134.1 | - | 2 | 562 | 6834018 | ||
| U62391.1 | - | 2 | 212 | |||