Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PSAP SIRNA Assays ›
          Gene Symbol:
          Psap
          Gene Name:
          prosaposin
          Gene Aliases:
          AI037048, SGP-1
          Chromosome Location:
          Chr.10: 60277628 - 60302600 on Build GRCm38
          UniGene ID:
          Mm.277498
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68776

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Psap
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146120.1 NP_001139592.1 3, 4 372
          NM_001146121.1 NP_001139593.1 3, 4 372
          NM_001146122.1 NP_001139594.1 3, 4 372
          NM_001146123.1 NP_001139595.1 3, 4 372
          NM_001146124.1 NP_001139596.1 3, 4 288
          NM_011179.3 NP_035309.3 3, 4 372
          GenBank mRNA AK088369.1 - 3, 4 286
          AK089998.1 - 3, 4 290
          AK093881.1 - 3, 4 286
          AK135637.1 - 3, 4 286
          AK145607.1 - 3, 4 288
          AK145802.1 - 3, 4 287
          AK148150.1 - 3, 4 294
          AK148152.1 - 3, 4 313
          AK148545.1 - 3, 4 288
          AK149785.1 - 3, 4 289
          AK150331.1 - 3, 4 288
          AK150645.1 - 3, 4 286
          AK151034.1 - 3, 4 286
          AK151173.1 - 3, 4 287
          AK151230.1 - 3, 4 286
          AK151234.1 - 3, 4 288
          AK151251.1 - 3, 4 286
          AK151525.1 - 3, 4 288
          AK151529.1 - 3, 4 288
          AK151686.1 - 3, 4 322
          AK151852.1 - 3, 4 288
          AK151875.1 - 3, 4 286
          AK151891.1 - 3, 4 286
          AK151907.1 - 3, 4 288
          AK151943.1 - 3, 4 288
          AK152117.1 - 3, 4 288
          AK152126.1 - 3, 4 286
          AK152134.1 - 3, 4 288
          AK152203.1 - 3, 4 320
          AK152280.1 - 3, 4 286
          AK152319.1 - 3, 4 290
          AK152355.1 - 3, 4 288
          AK152408.1 - 3, 4 291
          AK152505.1 - 3, 4 288
          AK152557.1 - 3, 4 288
          AK152605.1 - 3, 4 288
          AK152819.1 - 3, 4 300
          AK153404.1 - 3, 4 286
          AK159108.1 - 3, 4 316
          AK159164.1 - 3, 4 286
          AK159167.1 - 3, 4 287
          AK159198.1 - 3, 4 286
          AK159221.1 - 3, 4 290
          AK159244.1 - 3, 4 286
          AK159266.1 - 3, 4 286
          AK159333.1 - 3, 4 286
          AK159341.1 - 3, 4 286
          AK159384.1 - 3, 4 286
          AK159414.1 - 3, 4 289
          AK159427.1 - 3, 4 288
          AK159451.1 - 3, 4 286
          AK159457.1 - 3, 4 300
          AK159463.1 - 3, 4 288
          AK159487.1 - 3, 4 286
          AK159492.1 - 3, 4 286
          AK159510.1 - 3, 4 286
          AK159528.1 - 3, 4 286
          AK159530.1 - 3, 4 286
          AK159589.1 - 3, 4 288
          AK159604.1 - 3, 4 286
          AK159622.1 - 2, 3 178
          AK159634.1 - 3, 4 286
          AK159638.1 - 3, 4 287
          AK159657.1 - 3, 4 286
          AK159664.1 - 3, 4 286
          AK159685.1 - 3, 4 286
          AK159692.1 - 3, 4 286
          AK159709.1 - 3, 4 286
          AK159720.1 - 3, 4 286
          AK159722.1 - 3, 4 286
          AK159728.1 - 3, 4 288
          AK159840.1 - 3, 4 286
          AK159861.1 - 3, 4 286
          AK159862.1 - 3, 4 288
          AK159877.1 - 3, 4 286
          AK159910.1 - 3, 4 286
          AK159915.1 - 3, 4 286
          AK159936.1 - 3, 4 288
          AK159982.1 - 3, 4 286
          AK159994.1 - 3, 4 286
          AK160015.1 - 3, 4 286
          AK160019.1 - 3, 4 288
          AK160021.1 - 3, 4 286
          AK160024.1 - 3, 4 286
          AK160046.1 - 3, 4 286
          AK160049.1 - 3, 4 288
          AK160073.1 - 3, 4 286
          AK160108.1 - 3, 4 286
          AK160877.1 - 3, 4 289
          AK161313.1 - 3, 4 303
          AK166700.1 - 3, 4 286
          AK166707.1 - 3, 4 285
          AK166744.1 - 3, 4 284
          AK166811.1 - 3, 4 286
          AK166849.1 - 3, 4 286
          AK166879.1 - 3, 4 288
          AK166909.1 - 3, 4 286
          AK166920.1 - 3, 4 286
          AK166946.1 - 3, 4 286
          AK166959.1 - 3, 4 286
          AK166984.1 - 3, 4 286
          AK167014.1 - 3, 4 286
          AK167025.1 - 3, 4 286
          AK167073.1 - 3, 4 286
          AK167099.1 - 3, 4 289
          AK167108.1 - 3, 4 286
          AK167111.1 - 3, 4 286
          AK167147.1 - 3, 4 286
          AK167198.1 - 3, 4 288
          AK167213.1 - 3, 4 286
          AK167237.1 - 3, 4 286
          AK167243.1 - 3, 4 287
          AK167252.1 - 3, 4 288
          AK167418.1 - 3, 4 288
          AK167719.1 - 3, 4 288
          AK167902.1 - 3, 4 356
          AK170416.1 - 3, 4 285
          AK180106.1 - 2, 3 142
          AK194829.1 - 2, 3 142
          AK197804.1 - 2, 3 142
          AK201095.1 - 2, 3 142
          AK203509.1 - 2, 3 142
          AK210181.1 - 2, 3 156
          BC036266.1 - 2, 3 152
          BC141089.1 - 3, 4 285 9055701
          BC145359.1 - 3, 4 285 9053917
          S36200.1 - 3, 4 262
          U27340.1 - 3, 4 286

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          sphingolipid metabolic process
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          regulation of lipid metabolic process
          regulation of MAPK cascade
          positive regulation of MAPK cascade
          developmental growth
          prostate gland growth
          epithelial cell differentiation involved in prostate gland development
          cellular response to organic substance
          negative regulation of hydrogen peroxide-induced cell death
          G-protein coupled receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0