Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPRC SIRNA Assays ›
          Gene Symbol:
          Ptprc
          Gene Name:
          protein tyrosine phosphatase, receptor type, C
          Gene Aliases:
          B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc
          Chromosome Location:
          Chr.1: 138062859 - 138175755 on Build GRCm38
          UniGene ID:
          Mm.391573
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 68893

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptprc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001111316.2 NP_001104786.2 8 761
          NM_001268286.1 NP_001255215.1 5 370
          NM_011210.4 NP_035340.3 6 442
          GenBank mRNA AK054056.1 - 6 443
          AK088875.1 - 1 1794
          AK154893.1 - 5 365
          BC028512.1 - 6 382
          BC131655.1 - 6 382
          M11934.1 - 5 272
          M14342.1 - 6 404
          M14343.1 - 2 935
          M92933.1 - 8 761

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of T cell mediated cytotoxicity
          positive regulation of T cell mediated cytotoxicity
          negative regulation of cytokine-mediated signaling pathway
          hematopoietic progenitor cell differentiation
          immunoglobulin biosynthetic process
          positive regulation of T cell mediated immunity
          regulation of humoral immune response mediated by circulating immunoglobulin
          positive regulation of humoral immune response mediated by circulating immunoglobulin
          negative regulation of protein kinase activity
          protein dephosphorylation
          negative regulation of cell adhesion involved in substrate-bound cell migration
          leukocyte cell-cell adhesion
          response to gamma radiation
          dephosphorylation
          B cell differentiation
          T cell differentiation
          positive regulation of B cell proliferation
          negative regulation of protein autophosphorylation
          heterotypic cell-cell adhesion
          peptidyl-tyrosine dephosphorylation
          T cell proliferation
          B cell proliferation
          positive regulation of T cell proliferation
          positive regulation of MAPK cascade
          cell cycle phase transition
          positive thymic T cell selection
          negative thymic T cell selection
          regulation of B cell differentiation
          positive regulation of T cell differentiation
          positive regulation of gamma-delta T cell differentiation
          positive regulation of alpha-beta T cell proliferation
          positive regulation of isotype switching to IgG isotypes
          bone marrow development
          stem cell development
          negative regulation of peptidyl-tyrosine phosphorylation
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          regulation of B cell receptor signaling pathway
          positive regulation of antigen receptor-mediated signaling pathway
          release of sequestered calcium ion into cytosol
          defense response to virus
          regulation of cell cycle
          positive regulation of hematopoietic stem cell migration
          positive regulation of stem cell proliferation
          regulation of extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          heparin binding
          hydrolase activity
          phosphatase activity
          protein kinase binding
          heparan sulfate proteoglycan binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0