Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNB2 SIRNA Assays ›
          Gene Symbol:
          CACNB2
          Gene Name:
          calcium voltage-gated channel auxiliary subunit beta 2
          Gene Aliases:
          CACNLB2, CAVB2, MYSB
          Chromosome Location:
          Chr.10: 18140560 - 18545881 on Build GRCh38
          UniGene ID:
          Hs.59093
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 6942

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000724.3 NP_000715.2 2 770
          NM_001167945.1 NP_001161417.1 3 603
          NM_201570.2 NP_963864.1 2 624
          NM_201571.3 NP_963865.2 3 603
          NM_201572.3 NP_963866.2 3 603
          NM_201590.2 NP_963884.2 2 400
          NM_201593.2 NP_963887.2 3 380
          NM_201596.2 NP_963890.2 3 380
          NM_201597.2 NP_963891.1 3 380
          XM_005252588.3 XP_005252645.1 2 628
          XM_006717502.3 XP_006717565.1 2 433
          XM_011519659.2 XP_011517961.1 2 400
          XM_011519660.2 XP_011517962.1 2 985
          XM_017016625.1 XP_016872114.1 2 540
          GenBank mRNA AB208917.1 - 2 656
          AF137376.1 - 3 318
          AF137377.1 - 2 161
          AF285239.2 - 2 158
          AF423189.1 - 2 155
          AF423190.1 - 3 236
          AF423191.1 - 3 320
          AF423192.1 - 2 176
          AF465485.1 - 2 164
          AK293837.1 - 3 535
          AK293937.1 - 2 185
          AK295113.1 - 2 521
          AK296471.1 - 3 287
          AK299873.1 - 2 526
          AK307526.1 - 3 603
          AK307800.1 - 2 346
          AK310943.1 - 2 400
          AL119895.1 - 3 268
          AY393858.1 - 3 236
          AY393859.1 - 2 158
          AY393860.1 - 3 320
          AY393861.1 - 3 236
          AY393862.1 - 2 158
          AY393863.1 - 2 158
          AY675091.1 - 3 320
          BC136409.1 - 3 343 9020397
          DB235418.1 - 2 400
          S60415.1 - 3 380
          U95019.1 - 2 656

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001252533, NM_023116, BC109156, AK078220, FM872407, and others.

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chemical synaptic transmission
          neuromuscular junction development
          visual perception
          positive regulation of calcium ion transport
          cardiac conduction
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during AV node cell action potential
          regulation of heart rate by cardiac conduction
          establishment of protein localization to plasma membrane
          membrane depolarization during atrial cardiac muscle cell action potential
          regulation of voltage-gated calcium channel activity
          positive regulation of high voltage-gated calcium channel activity
          positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          voltage-gated calcium channel activity
          calcium channel regulator activity
          calcium channel activity
          protein binding
          high voltage-gated calcium channel activity
          protein kinase binding
          protein domain specific binding
          actin filament binding
          phosphoprotein binding
          voltage-gated calcium channel activity involved in cardiac muscle cell action potential
          voltage-gated calcium channel activity involved in AV node cell action potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0