Search
Search

Mlx| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159384.1 | NP_001152856.1 | 1, 2 | 86 | ||
| NM_001159385.1 | NP_001152857.1 | 1, 2 | 86 | |||
| NM_011550.3 | NP_035680.3 | 1, 2 | 248 | |||
| GenBank mRNA | AF213670.1 | - | 1, 2 | 50 | ||
| AF213671.1 | - | 1, 2 | 50 | |||
| AF213672.1 | - | 1, 2 | 212 | |||
| AK011478.1 | - | 1, 2 | 66 | |||
| AK142605.1 | - | 1, 2 | 52 | |||
| AK177451.1 | - | 40 | ||||
| BC031380.1 | - | 1, 2 | 228 | 5252695 | ||
| BC038004.1 | - | 1, 2 | 228 | |||
| CK620266.1 | - | 1, 2 | 90 | |||
| U43548.1 | - | 1, 2 | 208 | |||
| U43607.1 | - | 1, 2 | 219 | |||