Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC27A1 SIRNA Assays ›
          Gene Symbol:
          Slc27a1
          Gene Name:
          solute carrier family 27 (fatty acid transporter), member 1
          Gene Aliases:
          FATP1, Fatp
          Chromosome Location:
          Chr.8: 71568676 - 71587304 on Build GRCm38
          UniGene ID:
          Mm.38165
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 70672

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc27a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011977.3 NP_036107.1 13 1935
          GenBank mRNA AK040923.1 - 12 1874
          AK137700.1 - 7 1032
          AK153133.1 - 10 1676
          AK155185.1 - 11 1741
          AK164798.1 - 11 1714
          AK165571.1 - 10 1757
          BC028937.1 - 13 1935 5362790
          U15976.1 - 11 1730

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          medium-chain fatty acid transport
          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          phosphatidylethanolamine biosynthetic process
          phosphatidic acid biosynthetic process
          phosphatidylglycerol biosynthetic process
          phosphatidylcholine biosynthetic process
          phosphatidylserine biosynthetic process
          phosphatidylinositol biosynthetic process
          transport
          lipid transport
          metabolic process
          response to cold
          fatty acid transport
          long-chain fatty acid transport
          positive regulation of heat generation
          cardiolipin biosynthetic process
          response to insulin
          adiponectin-activated signaling pathway
          negative regulation of phospholipid biosynthetic process
          positive regulation of protein serine/threonine kinase activity
          nucleotide binding
          catalytic activity
          acyl-CoA ligase activity
          long-chain fatty acid-CoA ligase activity
          succinate-CoA ligase activity
          3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
          fatty acid transporter activity
          fatty acid ligase activity
          ligase activity
          3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
          2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
          benzoyl acetate-CoA ligase activity
          2,4-dichlorobenzoate-CoA ligase activity
          very long-chain fatty acid-CoA ligase activity
          pivalate-CoA ligase activity
          cyclopropanecarboxylate-CoA ligase activity
          adipate-CoA ligase activity
          citronellyl-CoA ligase activity
          mentha-1,3-dione-CoA ligase activity
          thiophene-2-carboxylate-CoA ligase activity
          2,4,4-trimethylpentanoate-CoA ligase activity
          cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
          trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
          protein homodimerization activity
          branched-chain acyl-CoA synthetase (ADP-forming) activity
          aryl-CoA synthetase (ADP-forming) activity
          3-hydroxypropionyl-CoA synthetase activity
          perillic acid:CoA ligase (ADP-forming) activity
          perillic acid:CoA ligase (AMP-forming) activity
          (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
          (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
          pristanate-CoA ligase activity
          malonyl-CoA synthetase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0