Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PNP SIRNA Assays ›
          Gene Symbol:
          Pnp Pnp2
          Gene Name:
          purine-nucleoside phosphorylase
          purine-nucleoside phosphorylase 2
          Gene Aliases:
          AL024301, AU015798, Np, Np-1, Np-2, Pnp1
          -
          Chromosome Location:
          Chr.14: 50944303 - 50953412 on Build GRCm38
          Chromosome Location:
          Chr.14: 50956141 - 50964749 on Build GRCm38
          UniGene ID:
          Mm.17932
          Mm.482171
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 71263

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pnp
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013632.4 NP_038660.1 2 397
          GenBank mRNA AK049817.1 - 2 176
          AK153451.1 - 2 399
          BC003788.1 - 2 169 3600212
          BC052679.1 - 2 148 30062908
          CT010254.1 - 1 117
          CT010316.1 - 1 117
          L11290.1 - 2 138
          L11291.1 - 2 138
          M84563.1 - 195
          U35374.1 - 1 117
          X56548.1 - 2 194
          Gene Symbol: Pnp2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001123371.2 NP_001116843.1 2 194
          GenBank mRNA AK008143.1 - 2 198

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase nucleotide kinase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          nucleobase-containing compound metabolic process
          inosine catabolic process
          deoxyinosine catabolic process
          deoxyguanosine catabolic process
          GTP biosynthetic process
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          nucleoside metabolic process
          response to gamma radiation
          urate biosynthetic process
          positive regulation of T cell proliferation
          purine nucleoside metabolic process
          purine-containing compound salvage
          positive regulation of B cell differentiation
          positive regulation of DNA repair
          dGTP metabolic process
          guanosine catabolic process
          positive regulation of alpha-beta T cell differentiation
          negative regulation of T cell apoptotic process
          biological_process
          nucleoside binding
          purine nucleobase binding
          catalytic activity
          purine-nucleoside phosphorylase activity
          drug binding
          transferase activity
          transferase activity, transferring glycosyl groups
          transferase activity, transferring pentosyl groups
          phosphate ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0