Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNA5 SIRNA Assays ›
Gene Symbol:
KCNA5
Gene Name:
potassium voltage-gated channel subfamily A member 5
Gene Aliases:
ATFB7, HCK1, HK2, HPCN1, KV1.5, PCN1
Chromosome Location:
Chr.12: 5043919 - 5046788 on Build GRCh38
UniGene ID:
Hs.150208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7191
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNA5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002234.3
NP_002225.2
1
2850
GenBank mRNA
M55513.1
-
1
2854
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
Notch signaling pathway
response to mechanical stimulus
response to organic substance
regulation of vasoconstriction
regulation of membrane potential
response to hydrogen peroxide
regulation of potassium ion transport
negative regulation of potassium ion transport
regulation of insulin secretion
protein homooligomerization
negative regulation of cytosolic calcium ion concentration
potassium ion homeostasis
response to hyperoxia
membrane hyperpolarization
regulation of atrial cardiac muscle cell membrane repolarization
potassium ion export
potassium ion transmembrane transport
atrial cardiac muscle cell action potential
membrane repolarization during bundle of His cell action potential
membrane repolarization during SA node cell action potential
regulation of heart rate by cardiac conduction
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of myoblast proliferation
receptor binding
delayed rectifier potassium channel activity
protein binding
outward rectifier potassium channel activity
potassium channel inhibitor activity
protein kinase binding
alpha-actinin binding
voltage-gated potassium channel activity involved in bundle of His cell action potential repolarization
voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization
voltage-gated potassium channel activity involved in SA node cell action potential repolarization
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0