Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7201
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
1, 2
563
NM_001161352.1
NP_001154824.1
1, 2
563
NM_001161353.1
NP_001154825.1
1, 2
563
NM_001271519.1
NP_001258448.1
1, 2
563
NM_001322829.1
NP_001309758.1
1, 2
563
NM_001322830.1
NP_001309759.1
1, 2
563
NM_001322832.1
NP_001309761.1
1, 2
563
NM_001322835.1
NP_001309764.1
1, 2
563
NM_001322836.1
NP_001309765.1
1, 2
563
NM_001322837.1
NP_001309766.1
1, 2
563
NM_002247.3
NP_002238.2
1, 2
563
XM_005269776.3
XP_005269833.1
1, 2
552
XM_005269778.1
XP_005269835.1
1, 2
552
XM_005269781.2
XP_005269838.1
1, 2
484
XM_005269787.3
XP_005269844.1
1, 2
552
XM_005269789.2
XP_005269846.1
1, 2
484
XM_005269792.2
XP_005269849.1
1, 2
484
XM_005269796.2
XP_005269853.1
1, 2
484
XM_006717826.2
XP_006717889.1
1, 2
484
XM_011539777.1
XP_011538079.1
1, 2
552
XM_011539778.2
XP_011538080.1
1, 2
484
XM_011539780.1
XP_011538082.1
1, 2
552
XM_011539783.1
XP_011538085.1
1, 2
552
XM_011539785.1
XP_011538087.1
1, 2
552
XM_017016215.1
XP_016871704.1
1, 2
552
XM_017016222.1
XP_016871711.1
1, 2
552
GenBank mRNA
AB113382.1
-
1, 2
328
AB113575.1
-
1, 2
314
AF025999.1
-
1, 2
314
AF118141.1
-
1, 2
191
AY040849.1
-
1, 2
328
BC137115.1
-
1, 2
466
9021112
BC137137.1
-
1, 2
466
9021134
BC144496.1
-
1, 2
466
9053023
U09384.1
-
1, 2
381
U11058.2
-
1, 2
277
U11717.1
-
1, 2
338
U13913.1
-
1, 2
464
U23767.1
-
1, 2
386
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0