Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNAB2 SIRNA Assays ›
Gene Symbol:
KCNAB2
Gene Name:
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases:
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
Chromosome Location:
Chr.1: 5992298 - 6101193 on Build GRCh38
UniGene ID:
Hs.440497
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7222
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNAB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199860.1
NP_001186789.1
6, 7
849
NM_001199861.1
NP_001186790.1
6, 7
833
NM_001199862.1
NP_001186791.1
5, 6
678
NM_001199863.1
NP_001186792.1
4, 5
241
NM_003636.3
NP_003627.1
6, 7
775
NM_172130.2
NP_742128.1
5, 6
733
XM_005263514.2
XP_005263571.1
5, 6
488
XM_011542321.2
XP_011540623.1
6, 7
505
XM_011542322.2
XP_011540624.1
6, 7
488
XM_017002618.1
XP_016858107.1
7, 8
1019
XM_017002619.1
XP_016858108.1
7, 8
655
XM_017002620.1
XP_016858109.1
5, 6
678
XM_017002621.1
XP_016858110.1
7, 8
1019
GenBank mRNA
AF029749.1
-
5, 6
290
AF044253.1
-
4, 5
248
AK124696.1
-
6, 7
854
AK131252.1
-
5, 6
678
AK289819.1
-
6, 7
439
AK290044.1
-
6, 7
411
AK312395.1
-
6, 7
411
AK312871.1
-
5, 6
413
AK315811.1
-
6, 7
425
AK315858.1
-
4, 5
241
BC110351.1
-
6, 7
468
5214271
BC126424.1
-
6, 7
383
8992140
BC130413.1
-
6, 7
383
40146444
BI907219.1
-
6, 7
468
CR542264.1
-
4, 5
248
CR542287.1
-
4, 5
248
U33429.1
-
5, 6
324
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
hematopoietic progenitor cell differentiation
neuromuscular process
oxidation-reduction process
NADPH oxidation
potassium ion transmembrane transport
regulation of potassium ion transmembrane transport
regulation of protein localization to cell surface
aldo-keto reductase (NADP) activity
voltage-gated potassium channel activity
potassium channel regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0