Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GJA5 SIRNA Assays ›
          Gene Symbol:
          GJA5
          Gene Name:
          gap junction protein alpha 5
          Gene Aliases:
          ATFB11, CX40
          Chromosome Location:
          Chr.1: 147756199 - 147781127 on Build GRCh38
          UniGene ID:
          Hs.447968
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7257

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GJA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005266.6 NP_005257.2 2 299
          NM_181703.3 NP_859054.1 2 296
          XM_005272951.4 XP_005273008.1 2 649
          XM_017001044.1 XP_016856533.1 3 2338
          GenBank mRNA BC013313.2 - 2 267 3844139
          BQ231177.1 - 2 275
          BQ924948.1 - 2 291
          BT019415.1 - 1 155
          BT019416.1 - 1 155
          BX377692.2 - 2 296
          DR003194.1 - 2 299
          KJ534844.1 - 1 155

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019280, XM_008761328, BC070935, M83092, AF021806.

          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          angiogenesis
          negative regulation of glomerular filtration
          outflow tract morphogenesis
          endothelium development
          cardiac conduction system development
          mitral valve development
          pulmonary valve formation
          ventricular septum development
          atrial septum development
          septum primum development
          atrial ventricular junction remodeling
          potassium ion transport
          cell communication by chemical coupling
          positive regulation of cell communication by chemical coupling
          gap junction assembly
          embryonic limb morphogenesis
          embryonic heart tube development
          foramen ovale closure
          negative regulation of blood pressure
          positive regulation of vasoconstriction
          positive regulation of vasodilation
          artery morphogenesis
          protein oligomerization
          transmembrane transport
          regulation of cardiac muscle contraction
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          ventricular septum morphogenesis
          atrial septum morphogenesis
          ventricular cardiac muscle cell action potential
          SA node cell action potential
          AV node cell to bundle of His cell communication by electrical coupling
          bundle of His cell to Purkinje myocyte communication by electrical coupling
          cell communication by electrical coupling involved in cardiac conduction
          regulation of heart rate by cardiac conduction
          regulation of AV node cell action potential
          regulation of bundle of His cell action potential
          regulation of Purkinje myocyte action potential
          regulation of atrial cardiac muscle cell action potential
          regulation of renin secretion into blood stream
          regulation of membrane depolarization during cardiac muscle cell action potential
          vasomotion
          gap junction hemi-channel activity
          connexin binding
          gap junction channel activity involved in cardiac conduction electrical coupling
          gap junction channel activity involved in AV node cell-bundle of His cell electrical coupling

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0