Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNAB2 SIRNA Assays ›
          Gene Symbol:
          KCNAB2
          Gene Name:
          potassium voltage-gated channel subfamily A regulatory beta subunit 2
          Gene Aliases:
          AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
          Chromosome Location:
          Chr.1: 5992298 - 6101193 on Build GRCh38
          UniGene ID:
          Hs.440497
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7317

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNAB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199860.1 NP_001186789.1 9 965
          NM_001199861.1 NP_001186790.1 9 949
          NM_001199862.1 NP_001186791.1 8 794
          NM_001199863.1 NP_001186792.1 7 357
          NM_003636.3 NP_003627.1 9 891
          NM_172130.2 NP_742128.1 8 849
          XM_005263514.2 XP_005263571.1 8 604
          XM_011542321.2 XP_011540623.1 9 621
          XM_011542322.2 XP_011540624.1 9 604
          XM_017002618.1 XP_016858107.1 10 1135
          XM_017002619.1 XP_016858108.1 10 771
          XM_017002620.1 XP_016858109.1 8 794
          XM_017002621.1 XP_016858110.1 10 1135
          GenBank mRNA AF029749.1 - 8 406
          AF044253.1 - 7 364
          AK124696.1 - 9 970
          AK131252.1 - 8 794
          AK289819.1 - 9 555
          AK290044.1 - 9 527
          AK312395.1 - 9 527
          AK312871.1 - 8 529
          AK315811.1 - 9 541
          AK315858.1 - 7 357
          BC110351.1 - 9 584 5214271
          BC126424.1 - 9 499 8992140
          BC130413.1 - 9 499 40146444
          BI907219.1 - 9 584
          CR542264.1 - 7 364
          CR542287.1 - 7 364
          U33429.1 - 8 440

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          neuromuscular process
          oxidation-reduction process
          NADPH oxidation
          potassium ion transmembrane transport
          regulation of potassium ion transmembrane transport
          regulation of protein localization to cell surface
          aldo-keto reductase (NADP) activity
          voltage-gated potassium channel activity
          potassium channel regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-sv4sw:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0