Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABCG2 SIRNA Assays ›
Gene Symbol:
ABCG2
Gene Name:
ATP binding cassette subfamily G member 2 (Junior blood group)
Gene Aliases:
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1
Chromosome Location:
Chr.4: 88090264 - 88231417 on Build GRCh38
UniGene ID:
Hs.480218
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
7338
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABCG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257386.1
NP_001244315.1
2
456
NM_004827.2
NP_004818.2
2
601
XM_005263354.3
XP_005263411.1
2
444
XM_005263355.3
XP_005263412.1
2
551
XM_005263356.3
XP_005263413.1
2
327
XM_011532420.2
XP_011530722.1
4
715
XM_017008852.1
XP_016864341.1
2
444
XM_017008853.1
XP_016864342.1
2
241
GenBank mRNA
AB051855.1
-
2
152
AB056867.1
-
1
108
AF098951.2
-
2
346
AF103796.1
-
2
312
AF463519.1
-
1
116
AK290000.1
-
2
172
AY017168.1
-
2
301
AY289766.1
-
2
308
AY333755.1
-
1
117
AY333756.1
-
1
117
BC021281.2
-
2
601
4908836
BC092408.1
-
2
456
6181905
BI598784.1
-
2
659
DQ459561.1
-
2
230
DQ459562.1
-
2
230
DQ459563.1
-
2
230
DQ459565.1
-
2
221
DQ459566.1
-
2
223
DQ459568.1
-
2
222
DQ459570.1
-
2
313
DQ459571.1
-
2
313
DQ459573.1
-
2
203
DQ459574.1
-
2
162
DQ459575.1
-
2
156
DQ459576.1
-
2
228
DQ459577.1
-
2
226
DQ459578.1
-
2
222
DQ459579.1
-
2
222
DQ459580.1
-
2
179
DQ459581.1
-
2
174
DQ459582.1
-
2
413
DQ459583.1
-
2
365
DQ459584.1
-
2
365
DQ459585.1
-
2
364
DQ459586.1
-
2
362
DQ459587.1
-
2
355
DQ459588.1
-
2
258
DQ459589.1
-
2
196
DQ459591.1
-
2
218
DQ459592.1
-
2
206
DQ459593.1
-
2
246
DQ459594.1
-
2
246
DQ459595.1
-
2
223
DQ459596.1
-
2
221
DQ459597.1
-
2
341
DQ459598.1
-
2
313
DQ459599.1
-
2
313
DQ459600.1
-
2
206
DQ459601.1
-
2
206
DQ459603.1
-
2
159
DQ459604.1
-
2
222
DQ459605.1
-
2
186
DQ459606.1
-
2
178
DQ459607.1
-
1
122
DQ459608.1
-
1
47
DQ459609.1
-
2
162
DQ459610.1
-
2
208
DQ459611.1
-
2
206
DQ459612.1
-
2
202
DQ459613.1
-
2
202
HQ386990.1
-
2
206
HQ386991.1
-
2
206
HQ386992.1
-
2
206
HQ386993.1
-
2
206
HQ386994.1
-
2
206
HQ386995.1
-
2
206
HQ386996.1
-
2
206
HQ386997.1
-
2
206
HQ386998.1
-
2
365
HQ386999.1
-
2
365
HQ387000.1
-
2
365
HQ387001.1
-
2
365
HQ387002.1
-
2
365
HQ387003.1
-
2
365
HQ387004.1
-
2
365
HQ387005.1
-
2
365
HQ387006.1
-
2
413
HQ387007.1
-
2
222
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
drug transmembrane transport
cellular iron ion homeostasis
response to iron ion
heme transport
response to drug
xenobiotic transport
urate metabolic process
drug export
response to folic acid
embryonic process involved in female pregnancy
cellular response to dexamethasone stimulus
transporter activity
protein binding
ATP binding
xenobiotic-transporting ATPase activity
heme transporter activity
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0