Search
Search

Aif1| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019467.2 | NP_062340.1 | 5 | 445 | ||
| GenBank mRNA | AB013745.1 | - | 3 | 172 | ||
| AF074959.1 | - | 3 | 183 | |||
| AK006184.1 | - | 5 | 445 | |||
| AK006562.1 | - | 5 | 446 | |||
| BC021539.1 | - | 3 | 313 | 5346103 | ||
| CT010265.1 | - | 3 | 131 | |||
| CT010385.1 | - | 3 | 131 | |||
| D86382.1 | - | 3 | 208 | |||
| EF101555.1 | - | 2 | 83 | |||