Search
Search

Lsp1| Gene Symbol: Lsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136071.2 | NP_001129543.1 | 2 | 251 | ||
| NM_001271508.1 | NP_001258437.1 | 2 | 618 | |||
| NM_001271509.1 | NP_001258438.1 | 2 | 251 | |||
| NM_001271510.1 | NP_001258439.1 | 2 | 251 | |||
| NM_001271523.1 | NP_001258452.1 | 2 | 450 | |||
| NM_019391.3 | NP_062264.1 | 2 | 618 | |||
| GenBank mRNA | AK003081.1 | - | 2 | 200 | ||
| AK031587.1 | - | 2 | 591 | |||
| AK075783.1 | - | 2 | 451 | |||
| BC003796.1 | - | 2 | 310 | 3488528 | ||
| BC044736.1 | - | 2 | 206 | |||
| D49691.1 | - | 2 | 255 | |||
| M89956.1 | - | 2 | 193 | |||
| M90316.1 | - | 2 | 617 | |||
| S74179.1 | - | 2 | 164 | |||