Search
Search

Rfk| Gene Symbol: Rfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019437.3 | NP_062310.1 | 4 | 2444 | ||
| GenBank mRNA | AK002806.1 | - | 4 | 2404 | ||
| AV285208.1 | - | 1 | 228 | |||
| BB758770.1 | - | 1 | 502 | |||
| BC033521.1 | - | 4 | 2346 | 4527980 | ||
| BC051021.1 | - | 4 | 2355 | 6315425 | ||