Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNMA1 SIRNA Assays ›
          Gene Symbol:
          KCNMA1
          Gene Name:
          potassium calcium-activated channel subfamily M alpha 1
          Gene Aliases:
          BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
          Chromosome Location:
          Chr.10: 76869601 - 77637819 on Build GRCh38
          UniGene ID:
          Hs.144795
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7391

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KCNMA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001014797.2 NP_001014797.1 14, 15 1921
          NM_001161352.1 NP_001154824.1 14, 15 1921
          NM_001161353.1 NP_001154825.1 14, 15 1921
          NM_001271518.1 NP_001258447.1 13, 14 1759
          NM_001271519.1 NP_001258448.1 14, 15 1921
          NM_001322829.1 NP_001309758.1 14, 15 1921
          NM_001322830.1 NP_001309759.1 14, 15 1921
          NM_001322832.1 NP_001309761.1 14, 15 1921
          NM_001322835.1 NP_001309764.1 14, 15 1921
          NM_001322836.1 NP_001309765.1 14, 15 1921
          NM_001322837.1 NP_001309766.1 14, 15 1921
          NM_001322838.1 NP_001309767.1 15, 16 1881
          NM_002247.3 NP_002238.2 14, 15 1921
          XM_005269776.3 XP_005269833.1 14, 15 1910
          XM_005269778.1 XP_005269835.1 14, 15 1910
          XM_005269781.2 XP_005269838.1 14, 15 1842
          XM_005269787.3 XP_005269844.1 14, 15 1910
          XM_005269789.2 XP_005269846.1 14, 15 1842
          XM_005269792.2 XP_005269849.1 14, 15 1842
          XM_005269796.2 XP_005269853.1 14, 15 1842
          XM_006717826.2 XP_006717889.1 14, 15 1842
          XM_011539773.1 XP_011538075.1 15, 16 2042
          XM_011539774.1 XP_011538076.1 15, 16 2042
          XM_011539775.1 XP_011538077.1 15, 16 2042
          XM_011539776.1 XP_011538078.1 15, 16 2042
          XM_011539777.1 XP_011538079.1 14, 15 1910
          XM_011539778.2 XP_011538080.1 14, 15 1842
          XM_011539779.1 XP_011538081.1 15, 16 2042
          XM_011539780.1 XP_011538082.1 14, 15 1910
          XM_011539781.2 XP_011538083.1 15, 16 2042
          XM_011539782.2 XP_011538084.1 15, 16 1994
          XM_011539783.1 XP_011538085.1 14, 15 1910
          XM_011539784.1 XP_011538086.1 15, 16 2042
          XM_011539785.1 XP_011538087.1 14, 15 1910
          XM_017016207.1 XP_016871696.1 15, 16 2042
          XM_017016208.1 XP_016871697.1 15, 16 2042
          XM_017016209.1 XP_016871698.1 15, 16 2042
          XM_017016210.1 XP_016871699.1 15, 16 2042
          XM_017016211.1 XP_016871700.1 15, 16 2042
          XM_017016212.1 XP_016871701.1 15, 16 2042
          XM_017016213.1 XP_016871702.1 15, 16 2042
          XM_017016214.1 XP_016871703.1 15, 16 2042
          XM_017016215.1 XP_016871704.1 14, 15 1910
          XM_017016216.1 XP_016871705.1 15, 16 2042
          XM_017016217.1 XP_016871706.1 15, 16 1994
          XM_017016218.1 XP_016871707.1 13, 14 1367
          XM_017016219.1 XP_016871708.1 15, 16 2042
          XM_017016222.1 XP_016871711.1 14, 15 1910
          XM_017016223.1 XP_016871712.1 12, 13 1219
          GenBank mRNA AB113382.1 - 14, 15 1686
          AB113575.1 - 14, 15 1672
          AB209488.1 - 13, 14 1327
          AB524033.1 - 13, 14 1582
          AF025999.1 - 14, 15 1672
          AF118141.1 - 14, 15 1549
          AK128392.1 - 5, 6 2177
          AK310379.1 - 15, 16 1881
          AY040849.1 - 14, 15 1686
          BC137115.1 - 14, 15 1824 9021112
          BC137137.1 - 14, 15 1824 9021134
          BC144496.1 - 14, 15 1824 9053023
          BX648925.1 - 9, 10 903
          U02632.1 - 7, 8 779
          U09384.1 - 14, 15 1739
          U11058.2 - 14, 15 1635
          U11717.1 - 14, 15 1696
          U13913.1 - 14, 15 1822
          U23767.1 - 14, 15 1744

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031828, AY344964, AY330292, AF135265, U40603, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001253365, NM_001253366, NM_001253378, NM_001253377, NM_001253367, and others.

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          potassium ion transport
          response to osmotic stress
          cellular potassium ion homeostasis
          response to carbon monoxide
          regulation of ion transmembrane transport
          regulation of membrane potential
          positive regulation of apoptotic process
          negative regulation of cell volume
          response to calcium ion
          micturition
          smooth muscle contraction involved in micturition
          potassium ion transmembrane transport
          actin binding
          voltage-gated potassium channel activity
          protein binding
          calcium-activated potassium channel activity
          metal ion binding
          large conductance calcium-activated potassium channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0