Search
Search

Kif21b| Gene Symbol: Kif21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039472.1 | NP_001034561.1 | 2 | 405 | ||
| GenBank mRNA | AF202893.1 | - | 2 | 383 | ||
| AK089400.1 | - | 2 | 353 | |||
| BY742264.1 | - | 2 | 375 | |||
| CJ102014.1 | - | 2 | 405 | |||