Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYP2E1 SIRNA Assays ›
          Gene Symbol:
          Cyp2e1
          Gene Name:
          cytochrome P450, family 2, subfamily e, polypeptide 1
          Gene Aliases:
          Cyp2e
          Chromosome Location:
          Chr.7: 140763832 - 140774981 on Build GRCm38
          UniGene ID:
          Mm.21758
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 75310

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cyp2e1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021282.2 NP_067257.1 1 180
          GenBank mRNA BC013451.1 - 1 180 4223797
          BC042693.1 - 1 159
          DQ838976.1 - 1 167
          L11650.1 - 1 273
          X62595.1 - 1 165

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          triglyceride metabolic process
          xenobiotic metabolic process
          steroid metabolic process
          monoterpenoid metabolic process
          drug metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          heterocycle metabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          3-(3-hydroxyphenyl)propionate hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          4-chlorobenzaldehyde oxidase activity
          3,5-xylenol methylhydroxylase activity
          phenylacetate hydroxylase activity
          4-nitrophenol 4-monooxygenase activity
          dimethyl sulfide monooxygenase activity
          alpha-pinene monooxygenase [NADH] activity
          phenanthrene 9,10-monooxygenase activity
          1-hydroxy-2-naphthoate hydroxylase activity
          toluene 4-monooxygenase activity
          xylene monooxygenase activity
          dibenzothiophene monooxygenase activity
          6-hydroxy-3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          chlorophenol 4-monooxygenase activity
          carbon disulfide oxygenase activity
          toluene 2-monooxygenase activity
          1-hydroxy-2-oxolimonene 1,2-monooxygenase activity
          phenanthrene 1,2-monooxygenase activity
          tetrahydrofuran hydroxylase activity
          styrene monooxygenase activity
          toluene-4-sulfonate monooxygenase activity
          toluene-sulfonate methyl-monooxygenase activity
          3-methyl-2-oxo-1,2-dihydroquinoline 6-monooxygenase activity
          2-hydroxy-phenylacetate hydroxylase activity
          2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA 1,2-monooxygenase activity
          phenanthrene 3,4-monooxygenase activity
          toluene 3-monooxygenase activity
          4-hydroxyphenylacetate,NADH:oxygen oxidoreductase (3-hydroxylating) activity
          limonene monooxygenase activity
          oxygen binding
          enzyme binding
          heme binding
          2-methylnaphthalene hydroxylase activity
          1-methylnaphthalene hydroxylase activity
          bisphenol A hydroxylase A activity
          salicylate 5-hydroxylase activity
          isobutylamine N-hydroxylase activity
          branched-chain dodecylbenzene sulfonate monooxygenase activity
          3-HSA hydroxylase activity
          4-hydroxypyridine-3-hydroxylase activity
          2-octaprenyl-3-methyl-6-methoxy-1,4-benzoquinol hydroxylase activity
          6-hydroxynicotinate 3-monooxygenase activity
          metal ion binding
          tocotrienol omega-hydroxylase activity
          thalianol hydroxylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0