Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STK4 SIRNA Assays ›
Gene Symbol:
Stk4
Gene Name:
serine/threonine kinase 4
Gene Aliases:
AI447339, AU020804, Kas-2, Mst1, Ysk3
Chromosome Location:
Chr.2: 164074109 - 164155524 on Build GRCm38
UniGene ID:
Mm.234472
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
75698
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Stk4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_021420.3
NP_067395.1
2
124
GenBank mRNA
AF271360.1
-
2
137
AK028838.1
-
2
126
BC054521.1
-
2
150
6409342
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
cell morphogenesis
patterning of blood vessels
neural tube formation
endocardium development
protein phosphorylation
apoptotic process
signal transduction
regulation of mitotic cell cycle
central nervous system development
negative regulation of cell proliferation
phosphorylation
peptidyl-serine phosphorylation
signal transduction by protein phosphorylation
keratinocyte differentiation
stress-activated protein kinase signaling cascade
positive regulation of protein binding
hippo signaling
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of fat cell differentiation
negative regulation of organ growth
protein autophosphorylation
primitive hemopoiesis
cell differentiation involved in embryonic placenta development
regulation of cell differentiation involved in embryonic placenta development
negative regulation of canonical Wnt signaling pathway
hepatocyte apoptotic process
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
transcription factor binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding
protein homodimerization activity
metal ion binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0