Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RYR2 SIRNA Assays ›
          Gene Symbol:
          Ryr2
          Gene Name:
          ryanodine receptor 2, cardiac
          Gene Aliases:
          9330127I20Rik, RYR-2
          Chromosome Location:
          Chr.13: 11553099 - 12106945 on Build GRCm38
          UniGene ID:
          Mm.239871
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 77357

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ryr2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_023868.2 NP_076357.2 2 628
          GenBank mRNA AF295105.1 - 2 129
          AK020368.1 - 2 371
          BC043140.1 - 2 628
          BC059061.1 - 2 628

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_001035, X98330.
          This siRNA also targets the following transcripts in Rat: NM_001191043, BC167757, AB204523.

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of heart rate
          embryonic heart tube morphogenesis
          left ventricular cardiac muscle tissue morphogenesis
          cardiac muscle hypertrophy
          detection of calcium ion
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          multicellular organism development
          positive regulation of heart rate
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          regulation of cardiac muscle contraction by calcium ion signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          response to muscle activity
          calcium-mediated signaling
          BMP signaling pathway
          response to caffeine
          response to magnesium ion
          ion transmembrane transport
          calcium-mediated signaling using intracellular calcium source
          response to muscle stretch
          release of sequestered calcium ion into cytosol
          positive regulation of sequestering of calcium ion
          regulation of cytosolic calcium ion concentration
          negative regulation of cytosolic calcium ion concentration
          response to calcium ion
          response to redox state
          transmembrane transport
          regulation of cardiac muscle contraction
          cardiac muscle contraction
          canonical Wnt signaling pathway
          cytosolic calcium ion transport
          calcium ion transport into cytosol
          sarcoplasmic reticulum calcium ion transport
          calcium ion transmembrane transport
          cellular response to caffeine
          manganese ion transmembrane transport
          cellular response to epinephrine stimulus
          establishment of protein localization to endoplasmic reticulum
          ventricular cardiac muscle cell action potential
          Purkinje myocyte to ventricular cardiac muscle cell signaling
          type B pancreatic cell apoptotic process
          positive regulation of the force of heart contraction
          regulation of AV node cell action potential
          regulation of SA node cell action potential
          regulation of atrial cardiac muscle cell action potential
          regulation of ventricular cardiac muscle cell action potential
          positive regulation of calcium-transporting ATPase activity
          ion channel activity
          ryanodine-sensitive calcium-release channel activity
          calcium channel activity
          calcium ion binding
          protein binding
          calmodulin binding
          calcium-release channel activity
          enzyme binding
          protein kinase binding
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          identical protein binding
          protein self-association
          suramin binding
          calcium-induced calcium release activity
          scaffold protein binding
          organic cyclic compound binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-f6wtj:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline