Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADA SIRNA Assays ›
          Gene Symbol:
          ADA
          Gene Name:
          adenosine deaminase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 44619519 - 44651758 on Build GRCh38
          UniGene ID:
          Hs.654536
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7755

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000022.3 NP_000013.2 4 390
          NM_001322051.1 NP_001308980.1 4 390
          GenBank mRNA AK223397.1 - 4 340
          BC007678.2 - 4 351 3629376
          BC040226.1 - 4 357 5198265
          BI601734.1 - 4 380
          HY102642.1 - 4 391
          KJ534760.1 - 4 239
          KU177892.1 - 4 239
          KU177893.1 - 4 239
          X02994.1 - 4 334

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          deaminase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          histamine secretion
          trophectodermal cell differentiation
          liver development
          placenta development
          germinal center B cell differentiation
          positive regulation of germinal center formation
          negative regulation of leukocyte migration
          negative regulation of mature B cell apoptotic process
          adenosine catabolic process
          deoxyadenosine catabolic process
          aging
          purine ribonucleoside monophosphate biosynthetic process
          positive regulation of heart rate
          positive regulation of B cell proliferation
          purine nucleotide salvage
          positive regulation of T cell differentiation in thymus
          response to vitamin E
          regulation of cell-cell adhesion mediated by integrin
          T cell activation
          negative regulation of circadian sleep/wake cycle, non-REM sleep
          response to drug
          response to hydrogen peroxide
          purine-containing compound salvage
          hypoxanthine salvage
          response to morphine
          positive regulation of smooth muscle contraction
          dATP catabolic process
          inosine biosynthetic process
          xanthine biosynthetic process
          positive regulation of alpha-beta T cell differentiation
          lung alveolus development
          Peyer's patch development
          embryonic digestive tract development
          negative regulation of inflammatory response
          positive regulation of calcium-mediated signaling
          positive regulation of T cell receptor signaling pathway
          negative regulation of adenosine receptor signaling pathway
          negative regulation of penile erection
          negative regulation of thymocyte apoptotic process
          negative regulation of mucus secretion
          purine nucleoside binding
          adenosine deaminase activity
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0