Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASS1 SIRNA Assays ›
          Gene Symbol:
          ASS1
          Gene Name:
          argininosuccinate synthase 1
          Gene Aliases:
          ASS, CTLN1
          Chromosome Location:
          Chr.9: 130444707 - 130501274 on Build GRCh38
          UniGene ID:
          Hs.160786
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 7869
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ASS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000050.4 NP_000041.2 4 507
          NM_054012.3 NP_446464.1 3 445
          XM_005272200.3 XP_005272257.1 3 250
          XM_011518705.2 XP_011517007.1 4 391
          XM_017014729.1 XP_016870218.1 4 349
          GenBank mRNA AK027126.1 - 4 632
          AK293114.1 - 3 194
          AK304625.1 - 3 219
          AK315093.1 - 3 194
          AW672752.1 - 3 283
          BC009243.2 - 3 222 3010137
          BC013224.1 - 3 183
          BC021676.1 - 3 194 4042389
          BE207182.1 - 3 274
          BE794559.1 - 263
          BF305325.1 - 3 279
          BI603075.1 - 4 520
          BX403885.2 - 3 253
          KU177923.1 - 2 151
          KU177924.1 - 2 151
          KU177925.1 - 2 151
          KU177926.1 - 2 151
          X01630.1 - 3 226

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_013157, XM_006233911, M36708, BC063146, X12459.

          Matching Taqman Gene Expression Assay:


          Hs00540723_m1

          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          urea cycle
          citrulline metabolic process
          argininosuccinate metabolic process
          kidney development
          liver development
          arginine biosynthetic process
          aspartate metabolic process
          acute-phase response
          midgut development
          aging
          response to nutrient
          response to zinc ion
          response to mycotoxin
          response to estradiol
          response to drug
          positive regulation of nitric oxide biosynthetic process
          response to growth hormone
          diaphragm development
          response to fatty acid
          cellular response to lipopolysaccharide
          cellular response to amino acid stimulus
          cellular response to ammonium ion
          cellular response to cAMP
          cellular response to interferon-gamma
          cellular response to tumor necrosis factor
          cellular response to glucagon stimulus
          cellular response to oleic acid
          cellular response to amine stimulus
          cellular response to laminar fluid shear stress
          cellular response to dexamethasone stimulus
          negative regulation of leukocyte cell-cell adhesion
          argininosuccinate synthase activity
          protein binding
          ATP binding
          toxic substance binding
          amino acid binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0