Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STK4 SIRNA Assays ›
          Gene Symbol:
          STK4
          Gene Name:
          serine/threonine kinase 4
          Gene Aliases:
          KRS2, MST1, YSK3
          Chromosome Location:
          Chr.20: 44966474 - 45079977 on Build GRCh38
          UniGene ID:
          Hs.472838
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 796

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STK4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006282.3 NP_006273.1 2, 3 218
          XM_005260530.2 XP_005260587.1 2, 3 186
          XM_005260531.3 XP_005260588.1 2, 3 186
          XM_005260533.2 XP_005260590.1 2, 3 212
          XM_011529020.2 XP_011527322.1 2, 3 186
          XM_017028033.1 XP_016883522.1 2, 3 212
          GenBank mRNA AB209315.1 - 2, 3 195
          AK225326.1 - 177
          AK309169.1 - 2, 3 180
          AK315238.1 - 2, 3 156
          AL597960.1 - 2, 3 218
          BC015332.1 - 2, 3 171
          BC029511.1 - 2, 3 191
          BC058916.1 - 2, 3 142
          BC093768.1 - 2, 3 160 7939613
          CR933633.1 - 2, 3 166
          U18297.1 - 2, 3 142
          U60207.1 - 2, 3 165

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          patterning of blood vessels
          neural tube formation
          positive regulation of protein phosphorylation
          endocardium development
          protein phosphorylation
          apoptotic process
          signal transduction
          central nervous system development
          negative regulation of cell proliferation
          peptidyl-serine phosphorylation
          signal transduction by protein phosphorylation
          keratinocyte differentiation
          positive regulation of protein binding
          positive regulation of peptidyl-serine phosphorylation
          hippo signaling
          intracellular signal transduction
          positive regulation of apoptotic process
          positive regulation of fat cell differentiation
          negative regulation of organ growth
          protein autophosphorylation
          protein stabilization
          primitive hemopoiesis
          cell differentiation involved in embryonic placenta development
          regulation of cell differentiation involved in embryonic placenta development
          positive regulation of protein serine/threonine kinase activity
          negative regulation of canonical Wnt signaling pathway
          hepatocyte apoptotic process
          positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          transcription factor binding
          identical protein binding
          protein homodimerization activity
          protein serine/threonine kinase activator activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0