Search
Search

Fam45a| Gene Symbol: Fam45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167829.1 | NP_001161301.1 | 6 | 681 | ||
| NM_026437.3 | NP_080713.2 | 6 | 681 | |||
| GenBank mRNA | AK004643.1 | - | 5 | 601 | ||
| AK007867.1 | - | 6 | 641 | |||
| AK145769.1 | - | 6 | 647 | |||
| AK168474.1 | - | 5 | 595 | |||
| BC013514.1 | - | 6 | 640 | 4235293 | ||