Search
Search

Fam45a| Gene Symbol: Fam45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167829.1 | NP_001161301.1 | 9 | 1534 | ||
| NM_026437.3 | NP_080713.2 | 9 | 1555 | |||
| GenBank mRNA | AK004643.1 | - | 8 | 1475 | ||
| AK007867.1 | - | 9 | 1515 | |||
| AK145769.1 | - | 9 | 1521 | |||
| AK168474.1 | - | 8 | 1469 | |||
| BC013514.1 | - | 9 | 1516 | 4235293 | ||