Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UCKL1 SIRNA Assays ›
Gene Symbol:
Uckl1
Gene Name:
uridine-cytidine kinase 1-like 1
Gene Aliases:
1110007H10Rik, Urkl1
Chromosome Location:
Chr.2: 181569149 - 181581973 on Build GRCm38
UniGene ID:
Mm.232400
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
81134
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Uckl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_026765.3
NP_081041.2
2, 3
304
GenBank mRNA
AK144803.1
-
2, 3
310
BC016535.1
-
2, 3
304
2655873
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide kinase
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
phosphorylation
nucleotide binding
uridine kinase activity
ATP binding
phosphofructokinase activity
phosphorylase kinase regulator activity
cobinamide kinase activity
phytol kinase activity
kinase activity
phosphatidylinositol phosphate kinase activity
cyclin-dependent protein serine/threonine kinase regulator activity
transferase activity
phenol kinase activity
cyclin-dependent protein kinase activating kinase regulator activity
inositol tetrakisphosphate 2-kinase activity
heptose 7-phosphate kinase activity
aminoglycoside phosphotransferase activity
phosphatidylinositol 3-kinase activity
eukaryotic elongation factor-2 kinase regulator activity
eukaryotic elongation factor-2 kinase activator activity
LPPG:FO 2-phospho-L-lactate transferase activity
cytidine kinase activity
glycerate 2-kinase activity
(S)-lactate 2-kinase activity
phosphoserine:homoserine phosphotransferase activity
L-seryl-tRNA(Sec) kinase activity
phosphocholine transferase activity
polynucleotide 5'-hydroxyl-kinase activity
ATP-dependent polynucleotide kinase activity
GTP-dependent polynucleotide kinase activity
farnesol kinase activity
CTP:2-trans,-6-trans-farnesol kinase activity
geraniol kinase activity
geranylgeraniol kinase activity
CTP:geranylgeraniol kinase activity
prenol kinase activity
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
phosphatidylinositol bisphosphate kinase activity
inositol-3,4,6-trisphosphate 1-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0