Search
Search

Egln3| Gene Symbol: Egln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028133.2 | NP_082409.2 | 5 | 1522 | ||
| GenBank mRNA | AF421882.1 | - | 5 | 1424 | ||
| AJ310548.1 | - | 5 | 1503 | |||
| AK011502.1 | - | 4 | 826 | |||
| AK044787.1 | - | 5 | 1524 | |||
| AK165972.1 | - | 5 | 1453 | |||
| AK170732.1 | - | 5 | 1453 | |||
| BC044926.1 | - | 5 | 1432 | 5373240 | ||
| BC058278.1 | - | 5 | 1442 | 6528802 | ||