Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPS6KB1 SIRNA Assays ›
Gene Symbol:
Rps6kb1
Gene Name:
ribosomal protein S6 kinase, polypeptide 1
Gene Aliases:
2610318I15Rik, 4732464A07Rik, 70kDa, AA959758, AI256796, AI314060, S6K1, p70/85s6k, p70s6k
Chromosome Location:
Chr.11: 86498869 - 86544807 on Build GRCm38
UniGene ID:
Mm.394280
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
82860
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rps6kb1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_028259.4
NP_082535.1
10
1942
GenBank mRNA
AK085687.1
-
10
1911
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
behavioral fear response
regulation of translation
protein phosphorylation
apoptotic process
cell cycle
signal transduction
germ cell development
positive regulation of smooth muscle cell migration
phosphorylation
TOR signaling
response to insulin
cellular response to insulin stimulus
response to tumor necrosis factor
negative regulation of apoptotic process
protein kinase B signaling
positive regulation of translation
positive regulation of mitotic cell cycle
positive regulation of translational initiation
regulation of glucose import
negative regulation of insulin receptor signaling pathway
positive regulation of skeletal muscle tissue growth
positive regulation of smooth muscle cell proliferation
cellular response to growth factor stimulus
cellular response to organic cyclic compound
negative regulation of extrinsic apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
ribosomal protein S6 kinase activity
protein serine/threonine/tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
PDZ domain binding
peptide binding
protein phosphatase 2A binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0