Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATG7 SIRNA Assays ›
          Gene Symbol:
          Atg7
          Gene Name:
          autophagy related 7
          Gene Aliases:
          1810013K23Rik, Agp7, Apg7l, Atg7l, Gm21553
          Chromosome Location:
          Chr.6: 114643097 - 114860614 on Build GRCm38
          UniGene ID:
          Mm.275332
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 83222

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Atg7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001253717.1 NP_001240646.1 3 346
          NM_001253718.1 NP_001240647.1 3 316
          NM_028835.4 NP_083111.1 2 195
          GenBank mRNA AB079385.1 - 1 122
          AK007484.1 - 2 172
          AK146992.1 - 3 317
          AK165653.1 - 2 168
          AK166023.1 - 2 169
          AK166074.1 - 2 166
          AK170769.1 - 2 169
          AK171968.1 - 3 285
          AK172272.1 - 3 285
          BC058597.1 - 1 156 5368226
          BY239844.1 - 2 196
          CJ092493.1 - 3 331

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001012097, XM_006237033, XM_006237034, XM_008763239, BC082059.

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          liver development
          protein lipidation
          C-terminal protein lipidation
          cellular amino acid metabolic process
          transport
          autophagy
          cellular response to nitrogen starvation
          organelle organization
          mitochondrion organization
          adult walking behavior
          cellular response to starvation
          post-embryonic development
          positive regulation of autophagy
          protein transport
          macroautophagy
          positive regulation of macroautophagy
          cerebellar Purkinje cell layer development
          pyramidal neuron development
          central nervous system neuron axonogenesis
          cerebral cortex development
          protein catabolic process
          neuron projection development
          regulation of protein ubiquitination
          positive regulation of protein modification process
          protein modification by small protein conjugation
          piecemeal microautophagy of nucleus
          suppression by virus of host autophagy
          negative stranded viral RNA replication
          response to starvation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          late nucleophagy
          positive regulation of protein catabolic process
          neurological system process
          defense response to virus
          cardiac muscle cell development
          regulation of cell development
          negative regulation of cell death
          membrane organization
          positive regulation of mucus secretion
          cellular response to hyperoxia
          regulation of mitophagy
          regulation of hemopoiesis
          negative regulation of histone H4-K16 acetylation
          protein binding
          transcription factor binding
          small protein activating enzyme activity
          Atg12 activating enzyme activity
          Atg8 activating enzyme activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0