Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOL3 SIRNA Assays ›
          Gene Symbol:
          Nol3
          Gene Name:
          nucleolar protein 3 (apoptosis repressor with CARD domain)
          Gene Aliases:
          ARC, B430311C09Rik, MYC, NOP, Nop30
          Chromosome Location:
          Chr.8: 105275937 - 105281939 on Build GRCm38
          UniGene ID:
          Mm.475715
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 84431

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nol3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_030152.4 NP_084428.1 3, 4 819
          GenBank mRNA AK021023.1 - 3, 4 765
          AK053846.1 - 3, 4 820
          AK079536.1 - 3, 4 851
          AY459322.1 - 3, 4 827
          BC053971.1 - 3, 4 791

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          blood vessel remodeling
          response to ischemia
          mRNA splice site selection
          mRNA processing
          regulation of gene expression
          cardiac muscle cell apoptotic process
          negative regulation of striated muscle cell apoptotic process
          negative regulation of cardiac muscle cell apoptotic process
          negative regulation of tumor necrosis factor-mediated signaling pathway
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          negative regulation of muscle atrophy
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          response to injury involved in regulation of muscle adaptation
          regulation of NF-kappaB import into nucleus
          regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          myoblast differentiation
          smooth muscle cell proliferation
          protein oligomerization
          negative regulation of cytosolic calcium ion concentration
          negative regulation of mitochondrial calcium ion concentration
          negative regulation of necrotic cell death
          cellular response to hypoxia
          negative regulation of release of cytochrome c from mitochondria
          intrinsic apoptotic signaling pathway
          inhibition of cysteine-type endopeptidase activity
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of mitochondrial membrane permeability involved in apoptotic process
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of death-inducing signaling complex assembly
          negative regulation of hydrogen peroxide-induced cell death
          negative regulation of protein targeting to mitochondrion
          negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
          inhibition of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway
          receptor binding
          death receptor binding
          calcium ion binding
          protein binding
          kinase binding
          phosphatase binding
          death effector domain binding
          identical protein binding
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          metal ion binding
          caspase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0