Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGFBP3 SIRNA Assays ›
          Gene Symbol:
          IGFBP3
          Gene Name:
          insulin like growth factor binding protein 3
          Gene Aliases:
          BP-53, IBP3
          Chromosome Location:
          Chr.7: 45912245 - 45921272 on Build GRCh38
          UniGene ID:
          Hs.450230
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8658

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IGFBP3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000598.4 NP_000589.2 1, 2 543
          NM_001013398.1 NP_001013416.1 1, 2 561
          XM_017012152.1 XP_016867641.1 1, 2 542
          GenBank mRNA AK055632.1 - 2, 3 491
          AK056274.1 - 2, 3 415
          AK075483.1 - 2, 3 456
          AK091706.1 - 2, 3 286
          AK091745.1 - 3, 4 356
          AK092447.1 - 3, 4 371
          AK095408.1 - 2, 3 430
          AK095869.1 - 2, 3 387
          AK095900.1 - 2, 3 399
          AK095907.1 - 2, 3 409
          AK095974.1 - 237
          AK095991.1 - 2, 3 387
          AK097201.1 - 79
          AK122873.1 - 2, 3 474
          AK123735.1 - 1, 2 185
          AK125224.1 - 2, 3 252
          AK127537.1 - 2, 3 371
          AK297767.1 - 2, 3 498
          AK298143.1 - 2, 3 408
          AK315668.1 - 1, 2 543
          BC000013.2 - 1, 2 509 3506666
          BC018962.2 - 1, 2 489 3506929
          BC064987.1 - 1, 2 527 4850456
          DQ884396.1 - 2, 3 282
          DQ884397.1 - 2, 3 348
          DQ884398.1 - 2, 3 327
          M31159.1 - 1, 2 520
          X64875.1 - 1, 2 543

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          osteoblast differentiation
          negative regulation of protein phosphorylation
          protein phosphorylation
          apoptotic process
          negative regulation of cell proliferation
          negative regulation of signal transduction
          regulation of glucose metabolic process
          negative regulation of smooth muscle cell migration
          regulation of apoptotic process
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          regulation of insulin-like growth factor receptor signaling pathway
          positive regulation of insulin-like growth factor receptor signaling pathway
          cellular protein metabolic process
          type B pancreatic cell proliferation
          positive regulation of myoblast differentiation
          negative regulation of smooth muscle cell proliferation
          fibronectin binding
          protein binding
          insulin-like growth factor binding
          protein tyrosine phosphatase activator activity
          insulin-like growth factor I binding
          insulin-like growth factor II binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0