Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IGF1 SIRNA Assays ›
          Gene Symbol:
          IGF1
          Gene Name:
          insulin like growth factor 1
          Gene Aliases:
          IGF-I, IGFI, MGF
          Chromosome Location:
          Chr.12: 102395867 - 102481839 on Build GRCh38
          UniGene ID:
          Hs.160562
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 8767

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IGF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000618.4 NP_000609.1 3, 4 674
          NM_001111283.2 NP_001104753.1 4, 5 723
          NM_001111284.1 NP_001104754.1 3, 4 512
          XM_017019262.1 XP_016874751.1 4, 5 533
          XM_017019263.1 XP_016874752.1 3, 4 476
          GenBank mRNA AB209184.1 - 3, 4 625
          AK312231.1 - 3, 4 573
          BC152321.1 - 3, 4 555 40129266
          CR541861.1 - 3, 4 410
          M27544.1 - 3, 4 558
          M29644.1 - 3, 4 421
          M37484.1 - 3, 4 512
          U40870.1 - 3, 4 402
          X00173.1 - 3, 4 421
          X56773.1 - 3, 4 410
          X57025.1 - 3, 4 576

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010512, NM_001111276, NM_001111275, NM_001111274, CT010364, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          skeletal system development
          response to hypoxia
          cell activation
          platelet degranulation
          DNA replication
          regulation of translation
          movement of cell or subcellular component
          signal transduction
          Ras protein signal transduction
          muscle organ development
          female pregnancy
          aging
          response to nutrient
          memory
          cell proliferation
          positive regulation of cell proliferation
          response to heat
          glycolate metabolic process
          regulation of gene expression
          positive regulation of glycoprotein biosynthetic process
          positive regulation of cardiac muscle hypertrophy
          phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to activity
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          muscle hypertrophy
          myotube cell development
          positive regulation of smooth muscle cell migration
          proteoglycan biosynthetic process
          positive regulation of cell migration
          response to caffeine
          activation of protein kinase B activity
          response to lipopolysaccharide
          cellular response to insulin stimulus
          positive regulation of protein import into nucleus, translocation
          negative regulation of smooth muscle cell apoptotic process
          response to nicotine
          bone mineralization involved in bone maturation
          regulation of multicellular organism growth
          positive regulation of activated T cell proliferation
          positive regulation of tyrosine phosphorylation of Stat5 protein
          negative regulation of apoptotic process
          positive regulation of DNA binding
          positive regulation of MAPK cascade
          protein kinase B signaling
          positive regulation of insulin-like growth factor receptor signaling pathway
          cellular protein metabolic process
          myoblast differentiation
          positive regulation of fat cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          positive regulation of Ras protein signal transduction
          insulin-like growth factor receptor signaling pathway
          phosphatidylinositol-mediated signaling
          positive regulation of fibroblast proliferation
          positive regulation of smooth muscle cell proliferation
          positive regulation of epithelial cell proliferation
          positive regulation of protein secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          protein stabilization
          detection of mechanical stimulus involved in sensory perception
          response to glucocorticoid
          myoblast proliferation
          regulation of calcium ion transport
          negative regulation of oocyte development
          positive regulation of cell growth involved in cardiac muscle cell development
          ERK1 and ERK2 cascade
          positive regulation of calcineurin-NFAT signaling cascade
          cellular response to electrical stimulus
          cellular response to estradiol stimulus
          negative regulation of release of cytochrome c from mitochondria
          negative regulation of neuron death
          positive regulation of trophectodermal cell proliferation
          negative regulation of cholangiocyte apoptotic process
          positive regulation of transcription regulatory region DNA binding
          negative regulation of extrinsic apoptotic signaling pathway
          insulin receptor binding
          insulin-like growth factor receptor binding
          integrin binding
          hormone activity
          steroid binding
          protein binding
          growth factor activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0