Search
Search

Mavs| Gene Symbol: Mavs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206382.1 | NP_001193311.1 | 8 | 2588 | ||
| NM_001206383.1 | NP_001193312.1 | 6 | 2322 | |||
| NM_001206385.1 | NP_001193314.1 | 7 | 2497 | |||
| NM_144888.2 | NP_659137.1 | 7 | 2725 | |||
| GenBank mRNA | AK028421.1 | - | 7 | 2497 | ||
| AK085038.1 | - | 7 | 2687 | |||
| AK153748.1 | - | 8 | 2593 | |||
| BC020006.1 | - | 7 | 2457 | 4190175 | ||
| BC025825.1 | - | 7 | 2650 | 5100990 | ||
| BC031352.1 | - | 7 | 2634 | 5123637 | ||
| BC037391.1 | - | 6 | 2436 | 5361522 | ||