Search
Search

Kank2| Gene Symbol: Kank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145611.4 | NP_663586.3 | 3 | 817 | ||
| GenBank mRNA | AK031324.1 | - | 3 | 808 | ||
| AK045492.1 | - | 3 | 789 | |||
| AK049288.1 | - | 3 | 818 | |||
| BC006647.1 | - | 3 | 805 | |||
| BC010245.1 | - | 3 | 774 | 3710765 | ||