Search
Search

Mapre2| Gene Symbol: Mapre2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162941.1 | NP_001156413.1 | 3 | 252 | ||
| NM_001162942.1 | NP_001156414.1 | 3 | 195 | |||
| NM_153058.4 | NP_694698.3 | 2 | 303 | |||
| GenBank mRNA | AI325879.1 | - | 3 | 189 | ||
| AK028823.1 | - | 3 | 253 | |||
| AK038562.1 | - | 2 | 290 | |||
| AK040767.1 | - | 2 | 278 | |||
| AK045493.1 | - | 2 | 186 | |||
| AK045904.1 | - | 2 | 278 | |||
| AK046095.1 | - | 2 | 245 | |||
| AK046221.1 | - | 2 | 270 | |||
| AK148954.1 | - | 2 | 278 | |||
| AK168834.1 | - | 3 | 137 | |||
| BC025804.1 | - | 2 | 250 | 5101879 | ||
| BC027056.1 | - | 2 | 269 | 5361238 | ||
| BC028987.1 | - | 2 | 225 | 5362960 | ||
| BC035254.1 | - | 2 | 255 | 5363176 | ||