Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMK2D SIRNA Assays ›
          Gene Symbol:
          CAMK2D
          Gene Name:
          calcium/calmodulin dependent protein kinase II delta
          Gene Aliases:
          CAMKD
          Chromosome Location:
          Chr.4: 113451032 - 113761927 on Build GRCh38
          UniGene ID:
          Hs.144114
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 93

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMK2D
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001221.3 NP_001212.2 13, 14 1851
          NM_001321566.1 NP_001308495.1 14, 15 1893
          NM_001321567.1 NP_001308496.1 15, 16 1953
          NM_001321568.1 NP_001308497.1 15, 16 1926
          NM_001321569.1 NP_001308498.1 15, 16 1953
          NM_001321570.1 NP_001308499.1 15, 16 1953
          NM_001321571.1 NP_001308500.1 15, 16 1953
          NM_001321572.1 NP_001308501.1 15, 16 1953
          NM_001321573.1 NP_001308502.1 15, 16 1926
          NM_001321574.1 NP_001308503.1 13, 14 1848
          NM_001321575.1 NP_001308504.1 14, 15 1893
          NM_001321576.1 NP_001308505.1 13, 14 1848
          NM_001321577.1 NP_001308506.1 14, 15 1890
          NM_001321578.1 NP_001308507.1 13, 14 1820
          NM_001321579.1 NP_001308508.1 15, 16 1953
          NM_001321581.1 NP_001308510.1 15, 16 1994
          NM_001321583.1 NP_001308512.1 14, 15 1893
          NM_001321584.1 NP_001308513.1 14, 15 1890
          NM_001321585.1 NP_001308514.1 15, 16 1987
          NM_001321586.1 NP_001308515.1 15, 16 1950
          NM_001321587.1 NP_001308516.1 15, 16 1926
          NM_001321588.1 NP_001308517.1 14, 15 1890
          NM_001321589.1 NP_001308518.1 14, 15 1893
          NM_001321590.1 NP_001308519.1 14, 15 1893
          NM_001321591.1 NP_001308520.1 15, 16 1926
          NM_172114.1 NP_742112.1 14, 15 1893
          NM_172115.2 NP_742113.1 13, 14 1851
          NM_172127.2 NP_742125.1 13, 14 1851
          NM_172128.2 NP_742126.1 13, 14 1851
          XM_005263253.3 XP_005263310.1 15, 16 1768
          XM_005263254.3 XP_005263311.1 15, 16 1789
          XM_006714330.2 XP_006714393.1 13, 14 1690
          XM_011532292.2 XP_011530594.1 11, 12 803
          XM_017008672.1 XP_016864161.1 14, 15 1732
          XM_017008673.1 XP_016864162.1 14, 15 1729
          XM_017008674.1 XP_016864163.1 15, 16 1789
          XM_017008675.1 XP_016864164.1 16, 17 1893
          XM_017008676.1 XP_016864165.1 15, 16 1129
          XM_017008677.1 XP_016864166.1 14, 15 1729
          XM_017008678.1 XP_016864167.1 13, 14 1666
          GenBank mRNA AB209288.1 - 14, 15 1529
          AF071569.1 - 13, 14 992
          AK025368.1 - 2, 3 102
          AK055642.1 - 13, 14 1369
          AK294517.1 - 15, 16 1244
          BC032784.1 - 13, 14 1054 5178265
          EF139846.1 - 4, 5 179
          U50361.1 - 2, 3 89
          U73738.1 - 3, 4 131

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          MAPK cascade
          regulation of cell growth
          response to hypoxia
          regulation of the force of heart contraction
          regulation of membrane depolarization
          regulation of transcription from RNA polymerase II promoter
          protein phosphorylation
          calcium ion transport
          regulation of heart contraction
          regulation of G2/M transition of mitotic cell cycle
          positive regulation of cardiac muscle hypertrophy
          regulation of cell communication by electrical coupling
          positive regulation of cardiac muscle cell apoptotic process
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          positive regulation of smooth muscle cell migration
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cellular potassium ion homeostasis
          endoplasmic reticulum calcium ion homeostasis
          positive regulation of Rac protein signal transduction
          positive regulation of GTPase activity
          protein autophosphorylation
          positive regulation of smooth muscle cell proliferation
          protein oligomerization
          relaxation of cardiac muscle
          regulation of ryanodine-sensitive calcium-release channel activity
          interferon-gamma-mediated signaling pathway
          regulation of cellular localization
          cell growth involved in cardiac muscle cell development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          cardiac muscle cell contraction
          regulation of heart rate by cardiac conduction
          regulation of cardiac muscle cell action potential
          regulation of cardiac muscle cell action potential involved in regulation of contraction
          regulation of cellular response to heat
          regulation of histone deacetylase activity
          regulation of cell communication by electrical coupling involved in cardiac conduction
          regulation of relaxation of cardiac muscle
          negative regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          negative regulation of sodium ion transmembrane transporter activity
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          sodium channel inhibitor activity
          titin binding
          protein homodimerization activity
          ion channel binding
          nitric-oxide synthase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0