Search
Search

Trpt1| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153597.2 | NP_705825.2 | 4 | 776 | ||
| GenBank mRNA | AK038830.1 | - | 3 | 608 | ||
| AK048558.1 | - | 925 | ||||
| AK090183.1 | - | 3 | 1907 | |||
| AK141582.1 | - | 563 | ||||
| BC027575.1 | - | 4 | 574 | 1364225 | ||